Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LY6K Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

6437
PRSI6437EA 435 EUR InStock
PRSI6437
Anti-LY6K Rabbit Polyclonal Antibody
Antikörper
LY6K Antibody: The lymphocyte antigen 6 complex locus K (LY6K), a member of cancer-testis antigen was initially identified as a molecular marker for head-and-neck squamous cell carcinoma as well a breast cancer. LY6K is a GPI-anchored membrane protein that is specifically associated with germ cell marker TEX101 and is strongly observed in testis, but only weakly in other tissues. LY6K mRNA was found to be upregulated in numerous bladder cancers due to gene amplification. Furthermore, knockdown experiments using LY6K siRNA reduced cell growth, migration and invasion in bladder carcinoma cell lines, suggesting that LY6K contributes to bladder cancer development.

Type:
Antigen: LY6K
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge