Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SEC62 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

6471
PRSI6471EA 435 EUR InStock
PRSI6471
Anti-SEC62 Rabbit Polyclonal Antibody
Antikörper
SEC62 Antibody: SEC62 is an integral membrane protein located in the rough endoplasmic reticulum (ER) and is part of the SEC61-SEC62-SEC63 complex that is the central component of the protein translocation apparatus of the ER membrane. It is speculated that SEC61-SEC62-SEC63 may perform post-translational protein translocation into the ER and might also perform the backward transport of ER proteins that are subject to the ubiquitin-proteasome-dependent degradation pathway. Silencing of this gene with RNAi dramatically reduced the migration and invasive potential of numerous tumor cell lines, suggesting that it may be an attractive target for therapy of various tumors.

Type:
Antigen: SEC62
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge