Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TRAP Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

6911
PRSI6911EA 435 EUR InStock
PRSI6911
Anti-TRAP Rabbit Polyclonal Antibody
Antikörper
TRAP Antibody: TRAP, also known as uteroferrin, is an iron containing, glycosylated, acid phosphatase. It is the most basic of the acid phosphatases and is the only form not inhibited by L(+)-tartrate. Along with the related protein ACP2, TRAP mediates the removal of mannose 6-phosphate residues from proteins targeted to lysosomes. TRAP is present in brain at low levels, but is expressed at a much higher level in liver.

Type:
Antigen: TRAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge