Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-AIMP1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

7513
PRSI7513EA 435 EUR InStock
PRSI7513
Anti-AIMP1 Rabbit Polyclonal Antibody
Antikörper
AIMP1 (Endothelial monocyte-activating polypeptide II, EMAP2) is a proinflammatory cytokine for monocytes and granulocytes. It is specifically induced by apoptosis and is involved in the control of angiogenesis, inflammation, and wound healing (1,2). AIMP1 was identified as one of three auxiliary factors of the mammalian aminoacyl tRNA synthetase (ARS) complex. It binds and facilitates the catalytic reaction of arginyl-tRNA synthetase. Recent studies show that CD23 plays an essential role in the AIMP1-induced immune response and might be a target in the treatment of inflammatory diseases.

AIMP1 antibody is affinity chromatography purified via peptide column.

AIMP1 antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: AIMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge