Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MS4A1 Mouse Monoclonal Antibody [clone: 2H7] (APC (Allophycocyanin))

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-009
PRSI76-009EA 343 EUR InStock
PRSI76-009
Anti-MS4A1 Mouse Monoclonal Antibody [clone: 2H7] (APC (Allophycocyanin))
Antikörper
The 2H7 monoclonal antibody specifically reacts with human CD20, a 33 to 37 kDa B-lymphocyte surface molecule. CD20 is an unglycosylated four-transmembrane phosphoprotein expressed by B cells in all stage of development, except the final plasma cells. It has been reported that the molecule is involved in B cell activation and has also been observed on a subset of circulating T lymphocytes.

Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: 2H7
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge