Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: Ber-H2] (FITC (Fluorescein Isothiocyanate))

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-012
PRSI76-012EA 488 EUR InStock
PRSI76-012
Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: Ber-H2] (FITC (Fluorescein Isothiocyanate))
Antikörper
The Ber-H2 monoclonal antibody specifically reacts with human CD30, a 120 kDa type I transmembrane glycoprotein of the tumor necrosis factor receptor superfamily (TNFR) also known as the Ki-1 antigen. CD30 can elicit signals leading to either activation or apoptosis through interaction with CD30 ligand (CD30L). It is expressed by a subset of extrafollicular activated B and T cells, lung macrophages, activated NK cells, lymphomas, and endothelial cells. It is highly expressed on Hodgkins and Reed-Sternberg cells.

Type: Primary
Antigen: TNFRSF8
Clonality: Monoclonal
Clone: Ber-H2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge