Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Emr1 Rat Monoclonal Antibody [clone: BM8.1] (VioletFluor® 450)

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-055
PRSI76-055EA 734 EUR InStock
PRSI76-055
Anti-Emr1 Rat Monoclonal Antibody [clone: BM8.1] (VioletFluor® 450)
Antikörper
The BM8.1 monoclonal antibody specifically binds to the Mouse125 kDa F4/80 antigen, expressed by most mature macrophages. F4/80 is a transmembrane protein used as a marker of macrophages, although it is also expressed on Kupffer and Langerhans cells. The expression of F4/80 antigen is upregulated on bone marrow cells stimulated in vitro with the macrophage colony stimulating factor. The F4/80 antigen is a requirement for the induction of CD8 T cells-mediated peripheral tolerance.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450 nm.

Type: Primary
Antigen: Emr1
Clonality: Monoclonal
Clone: BM8.1
Conjugation: VioletFluor® 450
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge