Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Thy1 Rat Monoclonal Antibody [clone: 30-H12] (PE (Phycoerythrin)-Cy7®)

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-074
PRSI76-074EA 488 EUR InStock
PRSI76-074
Anti-Thy1 Rat Monoclonal Antibody [clone: 30-H12] (PE (Phycoerythrin)-Cy7®)
Antikörper
The 30-H12 monoclonal antibody specifically binds to MouseCD90.2, an alloantigen known as Thy-1.2, expressed on thymocytes, mature T cells, epithelial cells, neurons, hematopoietic stem cells, and fibroblasts. CD90 is a membrane glycoprotein that regulates the adhesion and signal transduction in T lymphocytes, and the adhesion of thymocytes to thymic stroma. The interaction between 30-H12 and the antibody to the CD3/TCR complex upregulates thymocytes signal transduction and apoptosis and downregulates mature T cell proliferation. The 30-H12 antibody seems to be unable to cross-link with CD90.1.

Type: Primary
Antigen: Thy1
Clonality: Monoclonal
Clone: 30-H12
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge