Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Itgal Rat Monoclonal Antibody [clone: M17/4]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-138
PRSI76-138EA 409 EUR InStock
PRSI76-138
Anti-Itgal Rat Monoclonal Antibody [clone: M17/4]
Antikörper
The M17/4 monoclonal antibody specifically reacts with mouse CD11a, a 180 kDa glycoprotein. CD11a is the integrin alpha L chain that forms with CD18 the heterodimer molecule Lymphocyte Function-associated Antigen-1 (LFA-1). LFA-1 is expressed on all leukocytes and is involved in intercellular adhesions. The M17/4 antibody blocks many of the LFA-1 mediated cell interaction and in vivo it prolongs allograft survival, inhibits the autoimmune response, and reduces graft versus host reactions.

Type: Primary
Antigen: Itgal
Clonality: Monoclonal
Clone: M17/4
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge