Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SELP Mouse Monoclonal Antibody [clone: Psel.KO2.3] (PE (Phycoerythrin))

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-209
PRSI76-209EA 488 EUR InStock
PRSI76-209
Anti-SELP Mouse Monoclonal Antibody [clone: Psel.KO2.3] (PE (Phycoerythrin))
Antikörper
The Psel.KO2.3 monoclonal antibody specifically reacts with mouse CD62P (P-Selectin), a 140 kDa type I transmembrane glycoprotein also known as granule membrane protein 140 (GMP-140). Upon activation it is quickly transported to the plasma membrane from the alpha-granules of platelets and WEibel-Palade bodies of endothelial cells. It is essential to the functions of cell adhesion during inflammatory reactions and in the interaction of platelets with monocytes and neutrophils. The Psel.KO2.3 antibody does not block adhesion and binds to an epitope common to several species including mouse, human, and rat.

Type: Primary
Antigen: SELP
Clonality: Monoclonal
Clone: Psel.KO2.3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge