Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Cd4 Rat Monoclonal Antibody [clone: GK1.5]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-333
PRSI76-333EA 271 EUR InStock
PRSI76-333
Anti-Cd4 Rat Monoclonal Antibody [clone: GK1.5]
Antikörper
The GK1.5 monoclonal antibody specifically binds with the mouse CD4 molecule, also known as L3T4, a 55 kDa differentiation antigen which binds to the MHC class II. CD4 is expressed on most thymocytes, a subpopulation of mature T lymphocytes, dendritic cells, pluripotent hematopoietic stem cells, B cell precursors, and lymphoid precursors inside the thymus. It is also expressed on the mouse egg cell membrane, enhancing adhesion to MHC class II bearing sperm. By interaction with MHC class II on the surface of APC, CD4 initiates the development of T lymphocytes and helps the optimum functioning of mature T lymphocytes. The binding of the GK1.5 antibody blocks the binding of the Anti-Mouse CD4 RM4-5 antibody.

Type: Primary
Antigen: Cd4
Clonality: Monoclonal
Clone: GK1.5
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge