Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CD24 Mouse Monoclonal Antibody [clone: SN3] (FITC (Fluorescein Isothiocyanate))

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-398
PRSI76-398EA 488 EUR InStock
PRSI76-398
Anti-CD24 Mouse Monoclonal Antibody [clone: SN3] (FITC (Fluorescein Isothiocyanate))
Antikörper
The SN3 monoclonal antibody specifically reacts with human CD24, a 35 to 45 kDa molecule also known as the Heat Stable Antigen (HAS), Ly-52, Nectadrin. It can be used as a marker for distinguishing between lymphocyte developmental stages as its expression varies on T and B cells during differentiation. CD24 is also expressed on monocytes, dendritic cells, hematopoietic stem cells, epidermal Lagershans cells, and neurons.

Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: SN3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge