Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IL2RA Mouse Monoclonal Antibody [clone: BC96] (PE (Phycoerythrin)-Cy7®)

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-502
PRSI76-502EA 522 EUR InStock
PRSI76-502
Anti-IL2RA Mouse Monoclonal Antibody [clone: BC96] (PE (Phycoerythrin)-Cy7®)
Antikörper
The BC96 monoclonal antibody specifically reacts with the 55 kDa type I transmembrane glycoprotein known as the interleukin-2 receptor alpha (IL-2R alpha, also known as CD25). CD25 is expressed by the early progenitors of T and B lymphocytes lineage, and by activated mature T and B lymphocytes. CD25 is a low affinity interleukin-2 receptor, but its association with the IL-2 receptor beta chain (CD122) and the common gamma chain (CD 132) results in a high affinity IL-2R complex. CD25 plays an important role in B and T cell proliferation, differentiation, and activation.

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: BC96
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge