Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CEACAM8 Mouse Monoclonal Antibody [clone: G10F5] (FITC (Fluorescein Isothiocyanate))

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-591
PRSI76-591EA 522 EUR InStock
PRSI76-591
Anti-CEACAM8 Mouse Monoclonal Antibody [clone: G10F5] (FITC (Fluorescein Isothiocyanate))
Antikörper
The G10F5 monoclonal antibody specifically reacts with human CD66b, a 100 kDa glycosylphosphatidylinositol (GPI) linked protein in the granulocyte-specific activation family. CD66b is expressed on granulocytes, neutrophils, and eosinophils. It is reported that this molecule plays a role in regulating neutrophil activation and cellular adhesion. It was previously classified as CD67, but renamed CD66b in the Fifth HLDA Workshop.

Type: Primary
Antigen: CEACAM8
Clonality: Monoclonal
Clone: G10F5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge