Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Cd86 Rat Monoclonal Antibody [clone: GL-1 (GL1)] (PE (Phycoerythrin))

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-603
PRSI76-603EA 271 EUR InStock
PRSI76-603
Anti-Cd86 Rat Monoclonal Antibody [clone: GL-1 (GL1)] (PE (Phycoerythrin))
Antikörper
The GL-1 monoclonal antibody reacts specifically with the mouse B7-2 (also known as CD86), a costimulatory molecule expressed by B and T lymphocytes, macrophages (thioglycollate-induced from peritoneum), astrocytes and dendritic cells. The memory CD4+ T lymphocytes express CD86 (as mRNA and protein). CD86, and the B7-1 (CD 80) molecule, are ligands for CD152 and CD28, and influence the costimulatory interactions between lymphocytes B and T. B7-2 is also involved in the mouse natural killer cell-mediated cytotoxicity. The GL-1 antibody blocks the mixed lymphocyte reaction (MLR) and inhibits the T-cells stimulation by antigen-presenting cells. Mixtures of anti-B7-1 antibody and GL-1 were reported to inhibit the interaction between CD125 and its ligand with the in vivo priming of cytotoxic T lymphocytes.

Type: Primary
Antigen: Cd86
Clonality: Monoclonal
Clone: GL-1 (GL1)
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge