Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Cd27 Hamster Monoclonal Antibody [clone: LG.3A10]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-631
PRSI76-631EA 694 EUR InStock
PRSI76-631
Anti-Cd27 Hamster Monoclonal Antibody [clone: LG.3A10]
Antikörper
The LG.3A10 monoclonal antibody binds to CD27, 45 kDa molecule in the Tumor Necrosis Factor Receptor (TNFR) superfamily. It is expressed on memory B cells, NK cells, mature T cells, and a subset of thymocytes. Cd27 is involved in the interactions between B and T cells through its ligand CD70. The LG.3A10 antibody is cross-reactive with human, mouse and rat CD27.

Type: Primary
Antigen: Cd27
Clonality: Monoclonal
Clone: LG.3A10
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Human;Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge