Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Cd8b1 Rat Monoclonal Antibody [clone: H35-17.2] (APC (Allophycocyanin))

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-721
PRSI76-721EA 409 EUR InStock
PRSI76-721
Anti-Cd8b1 Rat Monoclonal Antibody [clone: H35-17.2] (APC (Allophycocyanin))
Antikörper
The H35-17.2 monoclonal antibody specifically reacts the mouse cd8 beta, a 32 kDa chain also known as Ly-3 and Lyt-3. CD8 is a T cell surface antigen co-receptor that interacts with MHC class I molecules on antigen presenting cells. It is expressed as a heterodimer with the alpha cd8 chain. The H35-17.2 antibody blocks T cell-mediated cytolysis of allogeneic lymphoma cells.

Type: Primary
Antigen: Cd8b1
Clonality: Monoclonal
Clone: H35-17.2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge