Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Cd28 Hamster Monoclonal Antibody [clone: 37.51]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-738
PRSI76-738EA 271 EUR InStock
PRSI76-738
Anti-Cd28 Hamster Monoclonal Antibody [clone: 37.51]
Antikörper
The 37.51 monoclonal antibody specifically reacts with mouse CD28 molecule, a 45 kDa homodimer. It is expressed on most thymocytes, at low density on most of the CD4+ and CD8+ peripheral T lymphocytes, and on the natural killer cells. It is additionally found on activated splenocytes, mast cells, and thymocytes. CD 28 is a costimulator of T lymphocytes and a ligand for CD80 (B7-1), and CD86 (B7-2). The CD28 signaling stimulates the expression of IL-2 and IL-2 receptor and the cytotoxicity of CD3-activated T lymphocytes.The 37.51 antibody stimulates the proliferation of T lymphocytes and natural killer cells and their respective cytokine production.

Type: Primary
Antigen: Cd28
Clonality: Monoclonal
Clone: 37.51
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge