Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-765
PRSI76-765EA 488 EUR InStock
PRSI76-765
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Antikörper
The HIB19 monoclonal antibody reacts with a human 95 kDa transmembrane glycoprotein known as CD19, which is expressed by B lymphocytes during all the developmental stages, except for the terminally differentiated plasma cells. CD19 is also expressed on follicular dendritic cells, and seems to ensure the regulation of B lymphocytes proliferation. CD19, CD21, CD81, MHC class II, and Leu13 can bind together and form a complex which associates with the B cell receptor (BCR) on the surface of B lymphocytes and facilitates the signal transduction.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge