Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CD59 Mouse Monoclonal Antibody [clone: OV9A2] (PE (Phycoerythrin))

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-812
PRSI76-812EA 316 EUR InStock
PRSI76-812
Anti-CD59 Mouse Monoclonal Antibody [clone: OV9A2] (PE (Phycoerythrin))
Antikörper
The OV9A2 monoclonal antibody specifically reacts with human CD59 (Protectin), a 19-25 kDa GPI-anchored glycoprotein. CD59 is expressed on hematopoietic and non-hematopoietic cells including lymphocytes, granulocytes, erythrocytes, monocytes and neurons. CD59 can inhibit complement cytolytic activity and the assembly of the membrane attack complex when bound to C8 and C9 complement components. It is involved in T cell activation and modulates T cell adhesion through interaction with CD2.

Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: OV9A2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge