Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CD69 Mouse Monoclonal Antibody [clone: FN50] (FITC (Fluorescein Isothiocyanate))

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-813
PRSI76-813EA 448 EUR InStock
PRSI76-813
Anti-CD69 Mouse Monoclonal Antibody [clone: FN50] (FITC (Fluorescein Isothiocyanate))
Antikörper
The FN50 monoclonal antibody specifically reacts with human CD69, the 27 to 33 kDa type II transmembrane protein also known as the very early activation antigen (VEA) or the activation inducer molecule (AIM). It is expressed as a disulfide-linked dimer on B cells, T cells, NK cells, platelets, eosinophils, and neutrophils. It increases in expression upon cell activation and seems to serve a role as a signaling receptor.

Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: FN50
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge