Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Eng Rat Monoclonal Antibody [clone: MJ7/18] (PE (Phycoerythrin))

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-828
PRSI76-828EA 488 EUR InStock
PRSI76-828
Anti-Eng Rat Monoclonal Antibody [clone: MJ7/18] (PE (Phycoerythrin))
Antikörper
The MJ7/18 monoclonal antibody specifically reacts with mouse CD105 (Endoglin), a 90 kDa homodimeric glycoprotein expressed on vascular endothelial cells. CD105 is a marker for tumor angiogenesis research by identifying proliferating endothelium. It is also suggested to be involved in embryonic angiogenesis and cellular adhesion.

Type: Primary
Antigen: Eng
Clonality: Monoclonal
Clone: MJ7/18
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge