Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Il7r Rat Monoclonal Antibody [clone: A7R34] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-918
PRSI76-918EA 271 EUR InStock
PRSI76-918
Anti-Il7r Rat Monoclonal Antibody [clone: A7R34] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Antikörper
The A7R34 monoclonal antibody specifically reacts with the mouse CD127, the alpha subunit of the IL-7 receptor, expressed by immature B lymphocytes in the bone marrow, CD4-/CD8-, CD4+, and CD8+ thymocytes, and by mature T lymphocytes at low levels. Mature T cells express CD117 at low levels in the periphery.The A7R34 antibody prevents the interaction between IL-7 and its receptor and the binding of mAb SB/199, which also recognizes mouse CD127.

Type: Primary
Antigen: Il7r
Clonality: Monoclonal
Clone: A7R34
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge