Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SDC1 Mouse Monoclonal Antibody [clone: DL-101] (PE (Phycoerythrin))

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-933
PRSI76-933EA 554 EUR InStock
PRSI76-933
Anti-SDC1 Mouse Monoclonal Antibody [clone: DL-101] (PE (Phycoerythrin))
Antikörper
The DL-101 monoclonal antibody specifically reacts with human CD138, a 85-92 kDa type I heparan sulfate proteoglycan. CD138 is known as Syndecan-1 and is expressed on pre-B cells, immature B cells, plasma cells, and malignant plasma cells, to the exclusion of mature circulating B cells. Syndecan-1 is also expressed on vascular smooth muscle, endothelial, neural, and embryonic mesenchymal cells. It is involved in cellular proliferation, cellular migration, and acts as an extracellular matrix receptor. Among the hematopoietic elements, CD138 is a useful marker for plasma cells.

Type: Primary
Antigen: SDC1
Clonality: Monoclonal
Clone: DL-101
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge