Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FASLG Mouse Monoclonal Antibody [clone: NOK-1] (APC (Allophycocyanin))

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-940
PRSI76-940EA 311 EUR InStock
PRSI76-940
Anti-FASLG Mouse Monoclonal Antibody [clone: NOK-1] (APC (Allophycocyanin))
Antikörper
The NOK-1 monoclonal antibody specifically reacts with human CD178, which is the CD95 or Fas ligand. CD178 is a TNF superfamily type II transmembrane glycoprotein expressed by activated T and NK cells and is involved in Fas-mediated apoptosis of lymphocytes. CD178 is also expressed by monocytes, neutrophils, granulocytes and the parenchymal cells of the retina and cornea. The NOK-1 antibody has been reported to bind to COOH-terminus of the Fas ligand in the region associated with Fas binding.

Type: Primary
Antigen: FASLG
Clonality: Monoclonal
Clone: NOK-1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge