Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Tnfsf4 Rat Monoclonal Antibody [clone: RM134L] (Biotin)

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-942
PRSI76-942EA 271 EUR InStock
PRSI76-942
Anti-Tnfsf4 Rat Monoclonal Antibody [clone: RM134L] (Biotin)
Antikörper
The RM134L monoclonal antibody specifically reacts with mouse CD252, a TNF/NGF superfamily member present on activated B lymphocytes and antigen-presenting cells. On activated B cells it enhances immunoglobulin secretion and cell proliferation. CD252 is also known as the OX-40 ligand and interacts with the OX-40 antigen found on the surface of activated T cells. The RML134L is reported to block the costimulatory activity of OX-40L.

Type: Primary
Antigen: Tnfsf4
Clonality: Monoclonal
Clone: RM134L
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge