Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NOTCH1 Mouse Monoclonal Antibody [clone: mN1A] (PE (Phycoerythrin))

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

76-998
PRSI76-998EA 488 EUR InStock
PRSI76-998
Anti-NOTCH1 Mouse Monoclonal Antibody [clone: mN1A] (PE (Phycoerythrin))
Antikörper
The mN1A monoclonal antibody reacts with the intracellular domain of human and mouse Notch1 with high affinity for the activated intracellular form and lower affinity for the full length heterodimeric Notch1 forms. It does not react with Notch2,3, or 4 and does not cross-react with rat thymocytes. The neurogenic locus homolog protein 1 (Notch1) is a trasmembrane receptor that regulates cell fate decisions. It is expressed in CD4-CD8- and CD4-CD8+ thymocytes, and plays a role in the regulation of myelopoiesis and lymphopoiesis.

Type: Primary
Antigen: NOTCH1
Clonality: Monoclonal
Clone: mN1A
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge