Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ARR3 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

7643
PRSI7643EA 435 EUR InStock
PRSI7643
Anti-ARR3 Rabbit Polyclonal Antibody
Antikörper
Arrestin 3 (ARR3) belongs to the Arrestin family of proteins that function as negative regulators of G protein-coupled receptor (GPCR) signaling (1,2). ARR3 binds the c-Jun N-terminal kinase 3 (JNK3) and scaffolds the apoptosis signal-regulating kinase 1 (ASK1)-MAPK kinase4-JNK3 cascade in a receptor-independent fashion, promoting JNK3 phosphorylation. ARR3 also mediates the internalization of the C-C chemokine receptor CCR7 upon the binding of its ligand CCL19, but not CCL21, suggesting that it helps mediate CCR7 receptor desensitization.

ARR3 antibody is affinity chromatography purified via peptide column.

ARR3 antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ARR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge