Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Ifng Rat Monoclonal Antibody [clone: XMG1.2] (Biotin)

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

77-042
PRSI77-042EA 271 EUR InStock
PRSI77-042
Anti-Ifng Rat Monoclonal Antibody [clone: XMG1.2] (Biotin)
Antikörper
The XMG1.2 is a neutralizing antibody that binds with the mouse Interferon-gamma (IFN-gamma) protein, a 15 to 17 kDa cytokine with significant antibacterial, antiviral, and antitumoral properties. When secreted by natural killer cells and by natural killer T lymphocytes, it regulates the immune response and supports adaptive immunity when produced by Th1 or CD8+ T lymphocytes. IFN-gamma plays an important role in the activation, the growth, and the differentiation of the macrophages, B and T lymphocytes, and natural killer cells. It interacts synergically with other cytokines, such as TNF-alpha, to inhibit proliferation of normal and transformed cells. IFN- gamma is the primary cytokine that defines Th-1 cells.The biological activity of IFN-gamma is not affected by glycosylation.

Type: Primary
Antigen: Ifng
Clonality: Monoclonal
Clone: XMG1.2
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge