Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IL4 Mouse Monoclonal Antibody [clone: 8D4-8] (PE (Phycoerythrin))

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

77-056
PRSI77-056EA 311 EUR InStock
PRSI77-056
Anti-IL4 Mouse Monoclonal Antibody [clone: 8D4-8] (PE (Phycoerythrin))
Antikörper
The 8D4-8 monoclonal antibody specifically reacts with human IL-4, expressed by the activated T cells and mast cells. IL-4 is species-specific and stimulates the proliferation and differentiation of B lymphocytes. It upregulates the expression of IgE receptors and class II MHC antigen. The 8D4-8 antibody neutralizes the IL-4 cytokine.

Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone: 8D4-8
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge