Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Ly6a Rat Monoclonal Antibody [clone: D7] (PE (Phycoerythrin)-Cy7®)

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

77-099
PRSI77-099EA 448 EUR InStock
PRSI77-099
Anti-Ly6a Rat Monoclonal Antibody [clone: D7] (PE (Phycoerythrin)-Cy7®)
Antikörper
The D7 monoclonal antibody specifically reacts with mouse Ly-6A/E (Sca-1), a 18 kDa GPI-linked surface protein. It is expressed on multipotent hematopoietic stem cells (HSC) and subsets of B and T cells and involved in B and T cell lymphocyte responses. Splenocytes from Ly-6.2 strains exhibit a larger number of D7-positive cells than those from the Ly-6.1 strains.

Type: Primary
Antigen: Ly6a
Clonality: Monoclonal
Clone: D7
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge