Human Recombinant CCL24 (from CHO cells)
Lieferant: PROSCIGesamtbewertung: 0
Bewertung: von 5
CCL24 (eotaxin-2) has been shown to induce chemotaxis of eosinophils, basophils and resting T lymphocytes but not monocytes and activated T lymphocytes. CCL24 is a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line. CCL24 interacts with chemokine receptor CCR3 to induce chemotaxis in eosinophils. Elevated levels of CCL24 has been seen in patients with aspirin-exacerbated respiratory disease (AERD), such as asthma.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.