Human recombinant G-CSF (from E. coli)
Lieferant: PROSCIGesamtbewertung: 0
Bewertung: von 5
Human Granulocyte-Colony-Stimulating Factor (G-CSF) is 20 kD glycoprotein containing internal disulfide bonds. It induces the survival, proliferation, and differentiation of neutrophilic granulocyte precursor cells and it functionally activates mature blood neutrophils. Among the family of colony-stimulating factors, G-CSF is the most potent inducer of terminal differentiation to granulocytes and macrophages of leukemic myeloid cell lines
- This recombinant protein can be used for biological assays. For research use only.
The synthesis of G-CSF can be induced by bacterial endotoxins, TNF, Interleukin-1, and GM-CSF. Prostaglandin E2 inhibits the synthesis of G-CSF. In epithelial, endothelial, and fibroblastic cells secretion of G-CSF is induced by Interleukin-17.