Human recombinant CXCL14 (from E. coli)
Lieferant: PROSCIGesamtbewertung: 0
Bewertung: von 5
Human Chemokine (C-X-C Motif) Ligand 14 (CXCL14) is constitutively expressed in certain normal tissues but is reduced or absent from many established tumor cell lines and human cancers. CXCL14 is known to be a chemoattractant for monocyte and dendritic cells. CXCL14 inhibits angiogenesis and exhibits antimicrobial activities. Mature human and mouse CXCL14 differ by only 2 amino acid residues.
- This recombinant protein can be used for biological assays. For research use only.