Drucken
Mouse recombinant Cadherin-3
Lieferant: PROSCI
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
92-591
PRSI92-591EA 513 EUR
PRSI92-591
Mouse recombinant Cadherin-3
Proteine und Peptide
Cadherin-3 (CDH3) is a single-pass type I membrane protein that belongs to the cadherin superfamily. CDH3 is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region, and a highly conserved cytoplasmic tail. CDH3 is expressed in some normal epithelial tissues and in some carcinoma cell lines. CDH3 preferentially interacts with themselves in a homophilic manner in connecting cells. CDH3 is involved in loss of heterosygosity events in breast and prostate cancer. Mutations in CDH3 have been associated with congential hypotrichosis with juvenile macular dystrophy.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.