Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TNFRSF21 Rat Monoclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

XA-1015
PRSIXA-1015EA 654 EUR InStock
PRSIXA-1015
Anti-TNFRSF21 Rat Monoclonal Antibody
Antikörper
DR6 antibody can be used to recognize human DR6 by Flow Cytometry. Method: Cells (5x105) were incubated on ice for 30 min in 50 μl FACS buffer (PBS, 5% FCS, 0.02% azide) containing 1 μg/mL of anti-DR6 MAb (Luke-1). After washing in FACS buffer, R-PE-conjugated antibody to rat IgG was added. Cells were incubated on ice for 30 min, washed and analyzed by flow cytometry. Method: 3 HEK 293T cells (5 x 105) were mock transfected (thin line) or transfected with an expression plasmid enabling surface expression of mDR6 (thick line). Cells were incubated on ice for 30 min in 50 μl FACS buffer (PBS, 5% Fetal calf serum, 0.02% azide) containing 1 μg/mL of Luke-1 antibody. After washing in FACS buffer, PE-conjugated antibody to rat IgG was added. Cells were incubated on ice for 30 min, washed and then analyzed by flow cytometry.

DR6 (monoclonal Luke-1) antibody was purified to ≥95% in SDS-PAGE.

PBS containing 0.02% sodium azide

DR6 antibody can be stored at 4°C, stable for one year.

Type:
Antigen: TNFRSF21
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge