Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-APMAP Mouse Monoclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

XA-1016
PRSIXA-1016EA 654 EUR InStock
PRSIXA-1016
Anti-APMAP Mouse Monoclonal Antibody
Antikörper
ACRP30 antibody can be used to recognize human DR6 by Flow Cytometry. Method: 0.5 μg of rhsACRP30headless was resolved by SDS-PAGE under reducing conditions, transferred to nitrocellulose and incubated with the Ne.Na mouse monoclonal antibody at a 1 μg/mL dilution for 1 hour. Proteins were visualized using a peroxidase-conjugated antibody to mouse IgG and a chemiluminescence detection system.

Adiponectin (monoclonal Ne.Na) antibody was purified from concentrated hybridoma tissue culture supernatant. Adiponectin (monoclonal Ne.Na) antibody was purified to ≥95% in SDS-PAGE.

PBS containing 0.02% sodium azide

Adiponectin antibody can be stored at 4°C, stable for one year.

Type:
Antigen: APMAP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge