Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IL16 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

XP-5170
PRSIXP-5170EA 509 EUR InStock
PRSIXP-5170
Anti-IL16 Rabbit Polyclonal Antibody
Antikörper
IL16 is a proinflammatory cytokine that is chemotactic for CD4+ T lymphocytes, monocytes, and eosinophils. IL16 is synthesized as a precursor molecule (pro IL16) of approximately 68 kDa and 631 amino acid residues lacking a signal peptide. The sequence and structure of IL16 is conserved across species. In addition to inducing chemotaxis, IL16 upregulates the IL2 receptor and upregulates HLADR expression. It also inhibits T cell receptor (TCR)/CD3 dependent activation, and suppresses HIV1 replication in vitro. Sources of IL16 include epithelial cells, mast cells, T lymphocytes (CD4+ and CD8+), macrophages, synovial fibroblasts, and eosinophils.

Anti-hIL-16 specific antibody was purified by affinity chromatography employing immobilized hIL-16 matrix.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: IL16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge