Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CCL13 Rabbit Polyclonal Antibody (Biotin)

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

XP-5224BT
PRSIXP-5224BTEA 566 EUR InStock
PRSIXP-5224BT
Anti-CCL13 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Human macrophage/monocyte chemotactic protein 4 (MCP4) is a member of the b-chemokine family (C-C) of Cytokines. Like Eotaxin, MCP4 is a powerful eosinophil chemo-attractant. Additionally, like MCP3, MCP4 elicits chemo-attraction of monocytes and T lymphocytes

Anti-hMCP-4 specific antibody was purified by affinity chromatography and then biotinylated.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CCL13
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge