Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ATF2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

XPS-1002
PRSIXPS-1002EA 686 EUR InStock
PRSIXPS-1002
Anti-ATF2 Rabbit Polyclonal Antibody
Antikörper
The activating transcription factor ATF-2 (also called CRE-BP1) binds to both AP-1 and CRE DNA response elements and is a member of the ATF/CREB family of leucine zipper proteins. ATF-2 has been implicated in the transcriptional regulation of a number of genes including cytokines, cell cycle control and apoptosis. Various forms of cellular stress, including inflammatory cytokines and UV irradiation, stimulate the transcriptional activity of ATF-2. Stress induced ATF-dependent transcription is dependent on phosphorylation of ATF. Serine 490 and serine 498 are novel phosphorylation sites on ATF that have recently been identified. In addition, ATF2 is particularly abundant in the brain and the ATF2 family of transcription factors is considered important substrates of signals upstream of the activation of genes associated with neuronal growth and differentiation. ATF expression has also been linked to the depression in humans.

The antibody also recognizes the phosphorylated 54k splice form of ATF2. Applications include Dot Blots (DB), Immunohistochemistry (IHC) and Western Blots (WB). When internally tested under ideal conditions the working dilutions were 1:1000 for DB, IHC and WB.

Affinity Purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

For long term storage –80°C is recommended, but shorter term storage at –20°C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.

Type:
Antigen: ATF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge