Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GRIN1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

XPS-2022
PRSIXPS-2022EA 686 EUR InStock
PRSIXPS-2022
Anti-GRIN1 Rabbit Polyclonal Antibody
Antikörper
The ion channels activated by glutamate are typically divided into two classes. Those that are sensitive to N-methyl-D-aspartate (NMDA) are designated NMDA receptors (NMDAR). The NMDAR plays an essential role in memory, neuronal development and it has also been implicated in several disorders of the central nervous system including Alzheimer’s disease, epilepsy and ischemic neuronal cell death. Increased membrane surface expression of the NR1 subunit of the receptor has been associated with synaptic plasticity. There are a number of different splice variants of the NR1. Differential splicing of three exons in the NR1 subunit generates up to eight NR1 splice variants and 7 of these have been identified in cDNA libraries. These exons encode a 21 amino acid N-terminal domain (N1) and adjacent sequences in the C-terminus (C1 and C2). Splicing out the C2 cassette eliminates the first stop codon and produces a new reading frame that generates a new sequence of 22 amino acids (C2'). Considerable attention has been focused on the distribution and expression of these splice variants that may affect the functional properties and regulation of the NMDAR.

Affinity Purified

Reconstitute in 50 μl PBS. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: Grin1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge