Thermal soft grip gloves, A143
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A143Y8RL A143Y8RM A143Y8RXL
PRTWA143Y8RLEA 3.4 EUR InStock
PRTWA143Y8RL PRTWA143Y8RM PRTWA143Y8RXL
Thermal soft grip gloves, A143
Handschuhe
Great for cold environments, this glove is dipped with foamed latex offering extreme breathability.
  • Warm 10 gauge acrylic liner for cold protection
  • Latex foam coating for excellent grip in wet and dry conditions
  • Palm dipped to increase dexterity and ventilation
  • Breathable seamless liner
  • Ergonomically designed for comfort and ease of use
Provides superior grip performance in wet or dry conditions.
Zertifizierungen: EN 388:2016 (2142X), EN 511 (X1X)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge