Tergsus Gloves
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A250ORRS
PRTWA250ORRSEA 7.8 EUR InStock
PRTWA250ORRS
Tergsus Gloves
Handschuhe
Durable goat grain leather glove with hook and loop fastening, cotton back, and reinforced fingertips. Suitable for light industrial applications.
  • Hook and loop strap for safe and secure fit
  • Suitable for light industrial applications
  • Leather palm greatly improves durability
  • Reinforced panels in high wear areas for maximum durability
Made from high-quality durable goat leather, with a cotton backing for breathability. Retail tag aids presentation for retail sales.
Colour: Orange
Zertifizierungen: EN ISO 21420: 2020, EN 388: 2016 + A1: 2018 3142X. CE certified.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge