Heat resistant welding gloves, A500
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A500RERXL
PRTWA500RERXLEA 9.6 EUR InStock
PRTWA500RERXL
Heat resistant welding gloves, A500
Handschuhe
14" cow split leather welding gauntlet with a one piece back and full cotton lining for comfort.
  • Maximum EN 407 burn behaviour resistance
  • Fully lined with soft and comfortable cotton
Ideal for welding and metal handling.
Zertifizierungen: EN 388:2016 (3243X), EN 407 (413244), EN 12477 Type A, AS/NZS 2161 .2, AS/NZS 2161 .3, AS/NZS 2161 .4, ANSI/ISEA 105: 2016 CUT Level A2
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge