Cut resistant gloves, A620
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A620GRRXL A620GRRS A620GRRXXL A620GRRM A620GRRXS A620GRRL
PRTWA620GRRXLEA 66 EUR InStock
PRTWA620GRRXL PRTWA620GRRS PRTWA620GRRXXL PRTWA620GRRM PRTWA620GRRXS PRTWA620GRRL
Cut resistant gloves, A620
Handschuhe
These seamless HPPE/glass fibre gloves with a palm PU coating provide dexterity, cut resistance and good grip.
  • Smooth PU coating for increased abrasion resistance
  • Palm dipped to increase dexterity and ventilation
  • 100% breathable seamless liner
  • 13 gauge liner for a perfect fit
  • Low linting construction for minimal contamination
Zertifizierungen: EN 388:2016 (4241BX), EN 407 (X1XXX).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge