Sleeve protector, cut resistant, A689
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A689GRR
PRTWA689GRREA 74.1 EUR InStock
PRTWA689GRR
Sleeve protector, cut resistant, A689
Arbeitskleidung
Maximum cut level five protective tubular sleeve provides cut and heat resistance for wrist and arm area. Thumb hole for secure fit. Machine washable.
  • A wide and increasing range of hand protection styles is available
  • Material: Glass fiber
  • Level D cut resistance
  • Superior cut and heat resistance
  • 14" (35 cm) cut resistant protective tubular sleeve
  • Thumb hole for secure fit
  • 13 gauge liner for a perfect fit
  • Retail bag which aids presentation for retail sales
Weight: 64 g
Zertifizierungen: AS/NZS 2161 .2, AS/NZS 2161 .3, AS/NZS 2161 .4, ANSI/ISEA 105 - 2016 CUT Level A4, EN 388:2016 - 4.X.4.X.D., EN 407 X.1.X.X.X.X., EN 420, CE food safe.
Bestellinformation: Sold singly.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge