Safety Shoes, Slip-On, Low, Cloro/Cloron S2 SRC
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
B0507BKR36 B0507BKR37 B0507BKR38 B0507BKR39 B0507BKR40 B0507WHR35 B0507WHR36 B0507BKR41 B0507BKR42 B0507BKR43 B0507BKR44 B0507BKR45 B0507BKR46 B0507BKR47 B0507BKR48 B0507WHR37 B0507WHR38 B0507WHR39 B0507WHR40 B0507WHR41 B0507WHR42 B0507WHR43 B0507WHR44 B0507WHR45 B0507WHR46 B0507WHR47 B0507WHR48
PRTWB0507BKR36EA 83.9 EUR InStock
PRTWB0507BKR36 PRTWB0507BKR37 PRTWB0507BKR38 PRTWB0507BKR39 PRTWB0507BKR40 PRTWB0507WHR35 PRTWB0507WHR36 PRTWB0507BKR41 PRTWB0507BKR42 PRTWB0507BKR43 PRTWB0507BKR44 PRTWB0507BKR45 PRTWB0507BKR46 PRTWB0507BKR47 PRTWB0507BKR48 PRTWB0507WHR37 PRTWB0507WHR38 PRTWB0507WHR39 PRTWB0507WHR40 PRTWB0507WHR41 PRTWB0507WHR42 PRTWB0507WHR43 PRTWB0507WHR44 PRTWB0507WHR45 PRTWB0507WHR46 PRTWB0507WHR47 PRTWB0507WHR48
Safety Shoes, Slip-On, Low, Cloro/Cloron S2 SRC
Schuhe und Stiefel
Anti-static footwear.
  • Water-resistant upper to prevent water penetration
  • 200 joules resistant toecap
  • Shock absorption in the heel region
  • Hydrocarbons-resistant
  • Men’s width
  • Women’s width
The Cloro/Cloron S2 SRC safety shoe features an orthopaedic, made-to-measure footbed that complies with european norms DGUV112-191.
Colour: Black or white
Zertifizierungen: EN ISO 20345: 2011 S2 SRC.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge