Safety Shoes, Velcro®, Mid, Base Cromo S2 SRC
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
B0508WHR36 B0508WHR37 B0508WHR38 B0508WHR39 B0508WHR40 B0508WHR41 B0508WHR42 B0508WHR43 B0508WHR44 B0508WHR45 B0508WHR46 B0508WHR47 B0508WHR48
PRTWB0508WHR36EA 85.3 EUR InStock
PRTWB0508WHR36 PRTWB0508WHR37 PRTWB0508WHR38 PRTWB0508WHR39 PRTWB0508WHR40 PRTWB0508WHR41 PRTWB0508WHR42 PRTWB0508WHR43 PRTWB0508WHR44 PRTWB0508WHR45 PRTWB0508WHR46 PRTWB0508WHR47 PRTWB0508WHR48
Safety Shoes, Velcro®, Mid, Base Cromo S2 SRC
Schuhe und Stiefel
The BASE Cromo safety shoe is available in both men's and women's width.
  • Anti-static footwear
  • Water-resistant upper to prevent water penetration
  • 200 joules resistant toe-cap
  • Shock absorption in the heel region
  • Hydrocarbons-resistant
These safety shoes feature an orthopaedic made to measure footbed, complying with european norms DGUV112-191.
Zertifizierungen: EN ISO 20345: 2011 S2 SRC.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge