Safety Shoes, Slip-On, Mid, Base Argo S2 SRC
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
B0510WHR35 B0510WHR36 B0510WHR37 B0510WHR38 B0510WHR39 B0510WHR40 B0510WHR41 B0510WHR42 B0510WHR43 B0510WHR44 B0510WHR45 B0510WHR46 B0510WHR47 B0510WHR48
PRTWB0510WHR35EA 72.2 EUR InStock
PRTWB0510WHR35 PRTWB0510WHR36 PRTWB0510WHR37 PRTWB0510WHR38 PRTWB0510WHR39 PRTWB0510WHR40 PRTWB0510WHR41 PRTWB0510WHR42 PRTWB0510WHR43 PRTWB0510WHR44 PRTWB0510WHR45 PRTWB0510WHR46 PRTWB0510WHR47 PRTWB0510WHR48
Safety Shoes, Slip-On, Mid, Base Argo S2 SRC
Schuhe und Stiefel
The BASE Argo safety shoes are available in both men's and women's width.
  • Anti-static footwear
  • Water-resistant upper to prevent water penetration
  • 200 joules resistant toe-cap
  • Shock absorption in the heel region
  • Hydrocarbons-resistant
These safety shoes feature an orthopaedic made to measure footbed, complying with european norms DGUV112-191.
Zertifizierungen: EN ISO 20345: 2011 S2 SRC.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge