BASE Footwear Grand Canyon/Tulum Safety Shoes, Lace-Up, Low, S1P, Black
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
B0666BKR35 B0666BKR36 B0666BKR37 B0666BKR38 B0666BKR39 B0666BKR40 B0666BKR41 B0666BKR42 B0666BKR43 B0666BKR44 B0666BKR45 B0666BKR46 B0666BKR47 B0666BKR48
PRTWB0666BKR35EA 140 EUR InStock
PRTWB0666BKR35 PRTWB0666BKR36 PRTWB0666BKR37 PRTWB0666BKR38 PRTWB0666BKR39 PRTWB0666BKR40 PRTWB0666BKR41 PRTWB0666BKR42 PRTWB0666BKR43 PRTWB0666BKR44 PRTWB0666BKR45 PRTWB0666BKR46 PRTWB0666BKR47 PRTWB0666BKR48
BASE Footwear Grand Canyon/Tulum Safety Shoes, Lace-Up, Low, S1P, Black
Schuhe und Stiefel
Grand Canyon/Tulum low safety shoes in black feature a Microfibre + Hi-Tech fabric upper, SmellStop anti-bacterial lining, Fresh’n Flex midsole, SlimCap toe cap, and AirTech + TPU-skin sole.
  • 200 joules resistant toe-cap
  • Puncture-resistant sole
  • Shock absorption in the heel region
  • Anti-static footwear
  • Electronic devices protection from electrostatic discharges
  • Hydrocarbons-resistant
  • Metal-free
Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces. Orthopaedic made to measure footbed, complying with european norms DGUV112-191.
Zertifizierungen: EN ISO 20345: 2011 S1P FO ESD SR; CEI EN 61340-4-3 S1P.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge